Alkaline detergent, powder, Suma Strip K9
Supplier: Diversey
Suma
DIVY7510952EA 296 EUR
DIVY7510952
Alkaline detergent, powder, Suma Strip K9
Cleaning Agents Detergents and Surface Cleansers
Highly alkaline powdered detergent for periodical removal of starch residues from dishes or heavily soiled surfaces in food areas.
  • Low foaming properties to improve cleaning efficiency
  • Inhibits scale build up