Weighing funnels, PP
Supplier: Merck
Z509329-1PAK Z409472-1PAK Z509353-1PAK Z509361-1PAK Z409480-1PAK Z509345-1PAK
SIALZ509329-1PAKEA 127 EUR
SIALZ509329-1PAK SIALZ409472-1PAK SIALZ509353-1PAK SIALZ509361-1PAK SIALZ409480-1PAK SIALZ509345-1PAK
Weighing funnels, PP
Boats Weighing Boats
These translucent weighing funnels are made of PP.
  • Residual sample can be rinsed to guarantee 100% transfer
  • Suitable for safe handling of hazardous materials
  • Very good chemical and temperature resistance
Pouring necks designed to fit volumetric flasks, micro-tubes or any narrow neck apparatus for spill free transfer.