Trimethylolpropane trimethacrylate inhibited, Technical Grade, Sigma-Aldrich®
Supplier: Merck
Synonyms: 2-Ethyl-2-hydroxymethyl-1,3-propanediol Trimethacrylate, TMPTMA, 2-Ethyl-2-((methacryloyloxy)methyl)propane-1,3-diyl bis(2-methylacrylate)
Stabilization: 250 ppm Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ)
M-Clarity™ quality level = MQ200
246840-100G 246840-500G
SIAL246840-100GEA 51.3 EUR
SIAL246840-100G SIAL246840-500G
Trimethylolpropane trimethacrylate inhibited, Technical Grade, Sigma-Aldrich®
Trimethylolpropane trimethacrylate
Trimethylolpropane trimethacrylate is a crosslinking agent.
Trimethylolpropane trimethacrylate is used in the preparation of macroporous poly (glycidyl methacrylate-co-trimethylolpropane trimethacrylate) materials with fine controlled porous properties.
Formula: C₁₈H₂₆O₆
MW: 338.40 g/mol
Density: 1.06 g/ml (25 °C) (lit.)
Flash Pt: >130 °C
MDL Number: MFCD00008588
CAS Number: 3290-92-4
UN: 3082
ADR: 9 III
Order Now
Specification Test Results
Infrared Spectrum Conforms to Structure
Purity (GC) Typically 90%
MEHQ as Inhibitor 150 - 350 ppm
Vapor Pressure <0.01 mmHg ( 20 °C)
Refractive Index n20/D 1.472 (lit.)
Vapor Density >1 (vs air)