Alkyltrimethylammonium bromide ≥95% (by TLC), white powder, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ200

M7635-100G M7635-1KG M7635-250G M7635-500G
SIALM7635-100GEA 234 EUR
SIALM7635-100G SIALM7635-1KG SIALM7635-250G SIALM7635-500G
Alkyltrimethylammonium bromide ≥95% (by TLC), white powder, Sigma-Aldrich®
Alkyltrimethylammonium bromide

Alkyltrimethylammonium bromide is a surfactant and can be used to study the forces and stability of foam films produced from soluble cationic surfactants.


Alkyltrimethylammonium bromide has been used in a study to assess the adsorption of sugar surfactants at the air/water interface. It has also been used in a study to investigate dynamical features in cationic micelles.


Predominantly C₁₄H₂₉N(CH₃)₃Br but also contains C₁₂ and C₁₆ homologs.


Soluble in water 5% (w/v).


Synonym: Cetrimide (per BP)

MDL Number: MFCD00130461
UN: 3263
ADR: 8 II

Order Now

Specification Test Results

Solubility (Turbidity) 50 mg/ml, H₂O Clear to Very Slightly Hazy
Purity (TLC) ≥95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR