Reusable half masks, Advantage 200 LS
Supplier: M S A
0303870500 0303870501 0303870502
111-2363EA 32.5 EUR
111-2363 111-2364 111-2365
Reusable half masks, Advantage 200 LS
Respirators Reusable Respirators Half Face Respirators
The Advantage 200 LS is a comfortable, efficient and economic half mask. It is ideal for applications where workers are exposed to various hazards from job to job, such as high concentrations of fumes, mists and gases. With its patented MultiFlex system the Advantage 200 LS offers maximum protection and high wearing comfort. The soft, pliable combination of rubber and plastic provides a customised and pressure-free fit, instantly conforming to the wearer’s unique facial characteristics.
  • Lightweight, cost-efficient
  • Thermoplastic rubber increases softness and decreases weight
  • Multiflex system equalises facial pressure
  • AnthroCurve conforms to each user's face
Applications: Asbestos abatement, welding, sanding and grinding, painting.