Accessories for goggle gear 500
Supplier: 3M
111-2371EA 18.1 EUR
111-2371
Accessories for goggle gear 500
Goggles
3M™ Goggle Gear™ transparent plastic prescription insert can be easily placed in the 3M™ Goggle Gear™ 500 (MMMAGG501SGAF-EU).
  • For polycarbonate prescription lenses up to + and - 6 dioptre
  • This lens is helpful for anyone who needs corrective vision to go along with eye protection
  • Insert available for 3M™ Goggle Gear, 500-Series Splash Goggles