Magnesium ≥99.8% (metals basis), rods, Length: 30 cm (12 in), Ø 3.3 cm (1.3 in)
Supplier: Thermo Fisher Scientific
10231.KG 10231.KJ 10231.KM
10231.KGEA 77.5 EUR
10231.KG 10231.KJ 10231.KM
Magnesium ≥99.8% (metals basis), rods, Length: 30 cm (12 in), Ø 3.3 cm (1.3 in)
Magnesium
Purity: 99.8% CAS No.: 7439-95-4 Physical Form: ≈450g/pc Odour: Odourless MDL No.: MFCD00085308
Formula: Mg
MW: 24.3 g/mol
Boiling Pt: 1107
Melting Pt: 651
Density: 1.74 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00085308
CAS Number: 7439-95-4
EINECS: 231-104-6
UN: 1869
ADR: 4.1 III
Merck Index: 13,05675