Magnesium ≥99.95% (metals basis), slug, Length: 6.35 mm (0.25 in), Ø 6.35 mm (0.25 in)
Supplier: Thermo Fisher Scientific
43298.09
43298.30
43298.18
43298.09EA
88.1
EUR
43298.09
43298.30
43298.18
Magnesium ≥99.95% (metals basis), slug, Length: 6.35 mm (0.25 in), Ø 6.35 mm (0.25 in)
Magnesium
Purity: 99.95%
CAS No.: 7439-95-4
Physical Form: ≈0.35g/slug
Odour: Odourless
MDL No.: MFCD00085308
Formula:
Mg Boiling Pt: 1107 °C (1013 hPa) Melting Pt: 651 °C Density: 1.74 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00085308 CAS Number: 7439-95-4 EINECS: 231-104-6 UN: 1869 ADR: 4.1,III Merck Index: 13,05675 |