Magnesium ≥99.95% (metals basis), slug, Length: 12.7 mm (0.50 in), Ø 6.35 mm (0.25 in)
Supplier: Thermo Fisher Scientific
43299.30 43299.09 43299.18
43299.30EA 1300 EUR
43299.30 43299.09 43299.18
Magnesium ≥99.95% (metals basis), slug, Length: 12.7 mm (0.50 in), Ø 6.35 mm (0.25 in)
Magnesium
Purity: 99.95% CAS No.: 7439-95-4 Physical Form: ≈0.7g/slug Odour: Odourless MDL No.: MFCD00085308
Formula: Mg
MW: 24.3 g/mol
Boiling Pt: 1107
Melting Pt: 651
Density: 1.74 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00085308
CAS Number: 7439-95-4
EINECS: 231-104-6
UN: 1869
ADR: 4.1 III
Merck Index: 13,05675