Magnesium, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
474754-25G 474754-5G
SIAL474754-25GEA 1690 EUR
SIAL474754-25G SIAL474754-5G
Magnesium, Sigma-Aldrich®
Magnesium
Formula: Mg
Boiling Pt: 1107 °C (1013 hPa)
Melting Pt: 651 °C
Density: 1.74 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00085308
CAS Number: 7439-95-4
EINECS: 231-104-6
UN: 1869
ADR: 4.1,III
Merck Index: 13,05675