Magnesium ≥99.8% (metals basis), powder -20+100 mesh
Supplier: Thermo Fisher Scientific
Danger
00869.30 00869.A1
00869.30EA 79.5 EUR
00869.30 00869.A1
Magnesium ≥99.8% (metals basis), powder -20+100 mesh
Magnesium
Purity: 99.8% CAS No.: 7439-95-4 Appearance: Silver grey Odour: Odourless UN No.: 1418 MDL No.: MFCD00085308
Formula: Mg
MW: 24.3 g/mol
Boiling Pt: 1107
Melting Pt: 651
Density: 1.74 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00085308
CAS Number: 7439-95-4
EINECS: 231-104-6
UN: 1418
ADR: 4.3 II
Merck Index: 13,05675