Strontium, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
460346-25G 460346-5G
SIAL460346-25GEA 2980 EUR
SIAL460346-25G SIAL460346-5G
Strontium, Sigma-Aldrich®
Strontium
Formula: Sr
MW: 87.62 g/mol
Boiling Pt: 1151
Melting Pt: 570…660 °C
Density: 2.38 g/cm³ (770 °C)
MDL Number: MFCD00134060
CAS Number: 7440-24-6
EINECS: 231-133-4
UN: 1393
ADR: 4.3 II
Merck Index: 12,08994