Aluminium ≥99.9999% (metals basis), slug, Puratronic®, Length: 3.175 mm (0.125 in), Ø 3.175 mm (0.125 in)
Supplier: Thermo Fisher Scientific
Synonyms: Aluminum
44326.09 44326.18
44326.09EA 149 EUR
44326.09 44326.18
Aluminium ≥99.9999% (metals basis), slug, Puratronic®, Length: 3.175 mm (0.125 in), Ø 3.175 mm (0.125 in)
Aluminium
Purity: 99.9999% CAS No.: 7429-90-5 Physical Form: ≈0.07g/slug Odour: Odourless MDL No.: MFCD00134029
Formula: Al
Boiling Pt: 2327 °C (1013 hPa)
Melting Pt: 660 °C
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321