Aluminium 99,9999 Metals basis, wire, Puratronic®, annealed, Ø 1.0 mm (0.04 in)
Supplier: Thermo Fisher Scientific
Synonyms: Aluminum
44334.G5 44334.G9
44334.G5EA 221 EUR
44334.G5 44334.G9
Aluminium 99,9999 Metals basis, wire, Puratronic®, annealed, Ø 1.0 mm (0.04 in)
Aluminium
Purity: 99.9999% CAS No.: 7429-90-5 Physical Form: ≈2.1g/m, Temper: soft Odour: Odourless MDL No.: MFCD00134029
Formula: Al
MW: 26.98 g/mol
Boiling Pt: 2327
Melting Pt: 660
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321