Aluminium ≥99.99% (metals basis), wire, Ø 0.38 mm (0.015 in)
Supplier: Thermo Fisher Scientific
Synonyms: Aluminum
45100.H2 45100.G6
45100.H2EA 154 EUR
45100.H2 45100.G6
Aluminium ≥99.99% (metals basis), wire, Ø 0.38 mm (0.015 in)
Aluminium
Purity: 99.99% CAS No.: 7429-90-5 Physical Form: ≈0.31g/m Odour: Odourless MDL No.: MFCD00134029
Formula: Al
Boiling Pt: 2327 °C (1013 hPa)
Melting Pt: 660 °C
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321