Gallium, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
263265-10G 263265-50G
SIAL263265-10GEA 119 EUR
SIAL263265-10G SIAL263265-50G
Gallium, Sigma-Aldrich®
Gallium
Formula: Ga
Boiling Pt: 2403 °C (1013 hPa)
Melting Pt: 29 °C
Density: 5.904 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134045
CAS Number: 7440-55-3
EINECS: 231-163-8
UN: 2803
ADR: 8,III
Merck Index: 12,04364