Indium ≥99.9999% (metals basis), shot, Puratronic® ≤ 5 mm (0.2 in)
Supplier: Thermo Fisher Scientific
10615.09
10615.18
10615.30
10615.09EA
176
EUR
10615.09
10615.18
10615.30
Indium ≥99.9999% (metals basis), shot, Puratronic® ≤ 5 mm (0.2 in)
Indium
Purity: 99.9999%
CAS No.: 7440-74-6
Odour: Odourless
MDL No.: MFCD00134048
Formula:
In MW: 114.82 g/mol Boiling Pt: 2000 Melting Pt: 156 Density: 7.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134048 CAS Number: 7440-74-6 EINECS: 231-180-0 Merck Index: 13,04971 |