Indium ≥99.99% (metals basis), foil, Thickness: 0.05 mm (0.002 in)
Supplier: Thermo Fisher Scientific
38371.FF 38371.GH 38371.FI
38371.FFEA 157 EUR
38371.FF 38371.GH 38371.FI
Indium ≥99.99% (metals basis), foil, Thickness: 0.05 mm (0.002 in)
Indium
Purity: 99.99% CAS No.: 7440-74-6 Physical Form: ≈0.23g/25x25mm Odour: Odourless MDL No.: MFCD00134048
Formula: In
Boiling Pt: 2000 °C (1013 hPa)
Melting Pt: 156 °C
Density: 7.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134048
CAS Number: 7440-74-6
EINECS: 231-180-0
Merck Index: 13,04971