Indium ≥99.99% (metals basis), rods, Ø 2.38 mm (0.094 in)
Supplier: Thermo Fisher Scientific
43388.RQ
43388.Z8
43388.RQEA
99.7
EUR
43388.RQ
43388.Z8
Indium ≥99.99% (metals basis), rods, Ø 2.38 mm (0.094 in)
Indium
Purity: 99.99%
CAS No.: 7440-74-6
Physical Form: ≈0.33g/cm
Odour: Odourless
MDL No.: MFCD00134048
Formula:
In MW: 114.82 g/mol Boiling Pt: 2000 Melting Pt: 156 Density: 7.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134048 CAS Number: 7440-74-6 EINECS: 231-180-0 Merck Index: 13,04971 |