Disinfectant, ClearKlens Oxifast, sterile
Supplier: Diversey
141-1424EA 427 EUR
141-1424
Disinfectant, ClearKlens Oxifast, sterile
Cleaning Agents Disinfectants
ClearKlens Oxifast is a ready to use solution based on peracetic acid that can be used for spraying or wiping.
  • For the disinfection of walls, floors, surfaces and process plants in the pharmaceutical industry
  • ClearKlens Oxifast sterile - double-packed
  • Ready to use sporicidal compound
  • Wide range of efficacy even with very short contact time: Bactericidal, yeasticidal, fungicidal, virucidal and sporicidal
  • Available in sterile and non sterile versions (different pack sizes and spray heads).
  • 0,2 μm filtered
  • Low environmental impact.
ClearKlens Oxifast is suitable for the disinfection of hard surfaces and the exterior cleaning of equipment in critical and non critical areas.
Certifikace: EN 1276, EN 1650, EN 13697, EN 13704 20 °C, EN 14476
Delivery information: Supplied with certificates of conformity (possibly also for sterility).
Upozornení: Use biocides safely. Always read the label and product information before use.