Indium, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
326607-10G 326607-50G
SIAL326607-10GEA 121 EUR
SIAL326607-10G SIAL326607-50G
Indium, Sigma-Aldrich®
Indium
Formula: In
Boiling Pt: 2000 °C (1013 hPa)
Melting Pt: 156 °C
Density: 7.3 g/cm³ (20 °C)
MDL Number: MFCD00134048
CAS Number: 7440-74-6
EINECS: 231-180-0
Merck Index: 13,04971