Indium, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
357308-1.8G
357308-7.2G
SIAL357308-1.8GEA
181
EUR
SIAL357308-1.8G
SIAL357308-7.2G
Indium, Sigma-Aldrich®
Indium
Formula:
In Boiling Pt: 2000 °C (1013 hPa) Melting Pt: 156 °C Density: 7.3 g/cm³ (20 °C) |
MDL Number:
MFCD00134048 CAS Number: 7440-74-6 EINECS: 231-180-0 Merck Index: 13,04971 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...