Indium, powder, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
264032-25G 264032-5G
SIAL264032-25GEA 737 EUR
SIAL264032-25G SIAL264032-5G
Indium, powder, Sigma-Aldrich®
Indium
Formula: In
Density: 7.31 g/cm³ (20 °C)
MDL Number: MFCD00134048
CAS Number: 7440-74-6
EINECS: 231-180-0
UN: 3089
ADR: 4.1,III