Carbon ≥99%, felt, Thickness: 1.27 cm (0.5 in)
Supplier: Thermo Scientific
Synonyms: Charcoal
42107.RF 42107.RR
42107.RFEA 144 EUR
42107.RF 42107.RR
Carbon ≥99%, felt, Thickness: 1.27 cm (0.5 in)
Carbon
Purity: 99.0% Physical Form: ≈9.86g/10x10cm MDL No.: MFCD00133992
Formula: C
MW: 12.01 g/mol
Boiling Pt: ∼4000 °C (1013 hPa)
Melting Pt: 3727 °C
Density: 3.65 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-955-3
Merck Index: 13,01818