Charcoal activated, powder, Norit® GSX, washed_H2SO4, activated_steam_acid wash
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Carbon activated, Activated carbon, Activated charcoal
L11860.22 L11860.36 L11860.0P L11860.A3
L11860.22EA 34 EUR
L11860.22 L11860.36 L11860.0P L11860.A3
Charcoal activated, powder, Norit® GSX, washed_H2SO4, activated_steam_acid wash
Charcoal activated
CAS No.: 7440-44-0 UN No.: 1362 MDL No.: MFCD00133992
Formula: C
MW: 12.01 g/mol
Melting Pt: 3550
Density: 2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3
UN: 1362
ADR: 4.2 III
REACH: 01-2119488894-16
Merck Index: 13,01818