Charcoal activated, pellets, Norit® ROW 0.8 mm, activated_steam
Supplier: Thermo Fisher Scientific
Synonyms: Carbon activated, Activated carbon, Activated charcoal
L16334.22 L16334.36 L16334.0P
L16334.22EA 38.9 EUR
L16334.22 L16334.36 L16334.0P
Charcoal activated, pellets, Norit® ROW 0.8 mm, activated_steam
Charcoal activated
CAS No.: 7440-44-0 MDL No.: MFCD00133992
Formula: C
MW: 12.01 g/mol
Melting Pt: 3550
Density: 2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3
UN: 1362
ADR: 4.2 III
REACH: 01-2119488894-16
Merck Index: 13,01818