Glassy carbon, rods, Ø 7 mm (0.28 in) Type 2
Supplier: Thermo Fisher Scientific
37999.DJ 37999.DM
37999.DJEA 403 EUR
37999.DJ 37999.DM
Glassy carbon, rods, Ø 7 mm (0.28 in) Type 2
Glassy carbon
Physical Form: ≈0.55g/cm MDL No.: MFCD00133992
Formula: C
MW: 12.01 g/mol
Melting Pt: 3652
Density: 1.95 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3
Merck Index: 13,01818