Glassy carbon, plate, Thickness: 1 mm (0.04 in) Type 1
Supplier: Thermo Fisher Scientific
38024.FI 38024.GH
38024.FIEA 408 EUR
38024.FI 38024.GH
Glassy carbon, plate, Thickness: 1 mm (0.04 in) Type 1
Glassy carbon
Formula: C
Melting Pt: 3652 °C
Density: 1.95 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3
Merck Index: 13,01818