Glassy carbon, plate, Thickness: 2 mm (0.08 in) Type 1
Supplier: Thermo Fisher Scientific
38025.FI 38025.GH
38025.FIEA 378 EUR
38025.FI 38025.GH
Glassy carbon, plate, Thickness: 2 mm (0.08 in) Type 1
Glassy carbon
Formula: C
Melting Pt: 3652 °C
Density: 1.95 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3
Merck Index: 13,01818