Graphite, plate, Thickness: 1.27 cm (0.5 in)
Supplier: Thermo Fisher Scientific
10132.RI 10132.RJ
10132.RIEA 119 EUR
10132.RI 10132.RJ
Graphite, plate, Thickness: 1.27 cm (0.5 in)
Graphite
MDL No.: MFCD00144065
Formula: C
MW: 12.01 g/mol
Melting Pt: 3827
Density: 2.09…2.23 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00144065
CAS Number: 7782-42-5
EINECS: 231-955-3
Merck Index: 12,04560