Graphite ≥99% (metals basis), rods, Length: 61 cm (24 in), Ø 1.27cm (0.5 in)
Supplier: Thermo Fisher Scientific
10134.KG 10134.KJ
10134.KGEA 52.9 EUR
10134.KG 10134.KJ
Graphite ≥99% (metals basis), rods, Length: 61 cm (24 in), Ø 1.27cm (0.5 in)
Graphite
Purity: 99% Physical Form: ≈130g/pc MDL No.: MFCD00144065
Formula: C
MW: 12.01 g/mol
Melting Pt: 3827
Density: 2.09…2.23 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00144065
CAS Number: 7782-42-5
EINECS: 231-955-3
Merck Index: 12,04560