Graphite natural 99,9995 Metals basis, powder -100 mesh
Supplier: Thermo Fisher Scientific
Warning
14735.TA 14735.TB 14735.TC 14735.TD
14735.TAEA 75.3 EUR
14735.TA 14735.TB 14735.TC 14735.TD
Graphite natural 99,9995 Metals basis, powder -100 mesh
Graphite
Formula: C
MW: 12.01 g/mol
Melting Pt: 3827
Density: 2.09…2.23 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00144065
CAS Number: 7782-42-5
EINECS: 231-955-3
UN: 1325
ADR: 4.1 III
Merck Index: 12,04560