Graphite natural 99,9995 Metals basis, powder 2-15 µm
Supplier: Thermo Fisher Scientific
Warning
14736.TA 14736.TB 14736.TC 14736.TD
14736.TAEA 101 EUR
14736.TA 14736.TB 14736.TC 14736.TD
Graphite natural 99,9995 Metals basis, powder 2-15 µm
Graphite
Formula: C
MW: 12.01 g/mol
Melting Pt: 3827
Density: 2.09…2.23 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00144065
CAS Number: 7782-42-5
EINECS: 231-955-3
UN: 1325
ADR: 4.1 III
Merck Index: 12,04560