Graphite (natural) ≥99.9995% (metals basis), powder -200 mesh
Supplier: Thermo Fisher Scientific
Warning
40795.TB 40795.TC 40795.TD
40795.TBEA 211 EUR
40795.TB 40795.TC 40795.TD
Graphite (natural) ≥99.9995% (metals basis), powder -200 mesh
Graphite
Formula: C
MW: 12.01 g/mol
Melting Pt: 3827
Density: 2.09…2.23 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00144065
CAS Number: 7782-42-5
EINECS: 231-955-3
UN: 1325
ADR: 4.1 III
Merck Index: 12,04560