Graphite 99,8 Metals basis, foil, Thickness: 0.4 mm (0.015 in)
Supplier: Thermo Fisher Scientific
42952.CG 42952.VA 42952.VU
42952.CGEA 142 EUR
42952.CG 42952.VA 42952.VU
Graphite 99,8 Metals basis, foil, Thickness: 0.4 mm (0.015 in)
Graphite
Purity: 99.8% MDL No.: MFCD00144065
Formula: C
MW: 12.01 g/mol
Melting Pt: 3827
Density: 2.09…2.23 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00144065
CAS Number: 7782-42-5
EINECS: 231-955-3
Merck Index: 12,04560