Graphite (natural) ≥99.9% (metals basis), soluble -10 mesh
Supplier: Thermo Fisher Scientific
43319.A1 43319.30 43319.18
43319.A1EA 263 EUR
43319.A1 43319.30 43319.18
Graphite (natural) ≥99.9% (metals basis), soluble -10 mesh
Graphite
Purity: 99.9% MDL No.: MFCD00144065
Formula: C
MW: 12.01 g/mol
Melting Pt: 3827
Density: 2.09…2.23 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00144065
CAS Number: 7782-42-5
EINECS: 231-955-3
Merck Index: 12,04560