Silicon ≥99.9% (metals basis), crystalline powder -100 mesh
Supplier: Thermo Fisher Scientific
Warning
00311.22 00311.36
00311.22EA 115 EUR
00311.22 00311.36
Silicon ≥99.9% (metals basis), crystalline powder -100 mesh
Silicon
Purity: 99.9% CAS No.: 7440-21-3 Appearance: White Odour: Odourless UN No.: 1346 MDL No.: MFCD00085311
Formula: Si
MW: 28.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00085311
CAS Number: 7440-21-3
EINECS: 231-130-8
UN: 1346
ADR: 4.1 III
Merck Index: 12,08635