Silicon ≥98.4% (metals basis), lumps 10 cm (3.9 in) & down
Supplier: Thermo Fisher Scientific
Warning
10335.A1 10335.A7
10335.A1EA 76.2 EUR
10335.A1 10335.A7
Silicon ≥98.4% (metals basis), lumps 10 cm (3.9 in) & down
Silicon
Purity: 98.4% CAS No.: 7440-21-3 Odour: Odourless MDL No.: MFCD00085311
Formula: Si
MW: 28.09 g/mol
Density: 2.34 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00085311
CAS Number: 7440-21-3
EINECS: 231-130-8
Merck Index: 12,08635