Silicon ≥99.999% (metals basis), lumps 15 cm (5.9 in) & down
Supplier: Thermo Scientific
Warning
39101.14 39101.22 39101.A1 39101.A4
39101.14EA 59.2 EUR
39101.14 39101.22 39101.A1 39101.A4
Silicon ≥99.999% (metals basis), lumps 15 cm (5.9 in) & down
Silicon
Purity: 99.999+% CAS No.: 7440-21-3 Odour: Odourless MDL No.: MFCD00085311
Formula: Si
MW: 28.09 g/mol
Boiling Pt: 2355 °C (1013 hPa)
Melting Pt: 1410 °C
Density: 2.34 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00085311
CAS Number: 7440-21-3
EINECS: 231-130-8
Merck Index: 12,08635