Silicon ≥99.999% (metals basis), rods, Ø 2.5cm (0.98 in)
Supplier: Thermo Fisher Scientific
Warning
39102.DM 39102.DO
39102.DMEA 534 EUR
39102.DM 39102.DO
Silicon ≥99.999% (metals basis), rods, Ø 2.5cm (0.98 in)
Silicon
Purity: 99.999% CAS No.: 7440-21-3 Physical Form: ≈11.43g/cm Odour: Odourless MDL No.: MFCD00085311
Formula: Si
MW: 28.09 g/mol
Density: 2.34 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00085311
CAS Number: 7440-21-3
EINECS: 231-130-8
Merck Index: 12,08635