Silicon ≥99.999% (metals basis), plate, Thickness: 6.35 mm (0.25 in)
Supplier: Thermo Fisher Scientific
Warning
39112.FF 39112.FI 39112.GH
39112.FFEA 133 EUR
39112.FF 39112.FI 39112.GH
Silicon ≥99.999% (metals basis), plate, Thickness: 6.35 mm (0.25 in)
Silicon
Purity: 99.999% CAS No.: 7440-21-3 Physical Form: ≈9.54g/25x25mm Odour: Odourless MDL No.: MFCD00085311
Formula: Si
MW: 28.09 g/mol
Density: 2.34 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00085311
CAS Number: 7440-21-3
EINECS: 231-130-8
Merck Index: 12,08635