Tin 99.5%, powder -100 mesh
Supplier: Thermo Fisher Scientific
Warning
317811000 317815000
ACRO317811000EA 44.9 EUR
ACRO317811000 ACRO317815000
Tin 99.5%, powder -100 mesh
Tin
Formula: Sn
Boiling Pt: 2270 °C (760 mmHg)
Melting Pt: 231.9 °C
Density: 7.3 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00133862
CAS Number: 7440-31-5
EINECS: 231-141-8
Merck Index: 12,09587
Order Now
Specification Test Results
Appearance Grey Powder
Total trace metal impurities ≤0.5 %