Tin ≥99.8% (metals basis), shot 8-20 mesh
Supplier: Thermo Scientific
10377.22 10377.36 10377.A3
10377.22EA 74 EUR
10377.22 10377.36 10377.A3
Tin ≥99.8% (metals basis), shot 8-20 mesh
Tin
Purity: 99.8% CAS No.: 7440-31-5 Odour: Odourless MDL No.: MFCD00133862
Formula: Sn
MW: 118.71 g/mol
Boiling Pt: 2270 °C (1013 hPa)
Melting Pt: 232 °C
Density: 7.31 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133862
CAS Number: 7440-31-5
EINECS: 231-141-8
Merck Index: 12,09587