Tin 99,8 Metals basis, powder -325 mesh
Supplier: Thermo Fisher Scientific
Warning
10379.22 10379.36 10379.A3
10379.22EA 59.1 EUR
10379.22 10379.36 10379.A3
Tin 99,8 Metals basis, powder -325 mesh
Tin
Purity: 99.8% CAS No.: 7440-31-5 Physical Form: Apparent density 3.2g/cm3 Odour: Odourless UN No.: 3089 MDL No.: MFCD00133862
Formula: Sn
MW: 118.71 g/mol
Boiling Pt: 2270
Melting Pt: 232
Density: 7.31 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133862
CAS Number: 7440-31-5
EINECS: 231-141-8
UN: 3089
ADR: 4.1 III
Merck Index: 12,09587