Tin 99,995 Metals basis, powder -100 mesh
Supplier: Thermo Fisher Scientific
11013.04 11013.09 11013.18 11013.30
11013.04EA 44.1 EUR
11013.04 11013.09 11013.18 11013.30
Tin 99,995 Metals basis, powder -100 mesh
Tin
Purity: 99.995% CAS No.: 7440-31-5 Appearance: Grey Odour: Odourless MDL No.: MFCD00133862
Formula: Sn
MW: 118.71 g/mol
Boiling Pt: 2270
Melting Pt: 232
Density: 7.31 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133862
CAS Number: 7440-31-5
EINECS: 231-141-8
Merck Index: 12,09587