Tin 99,5, mossy ACS
Supplier: Thermo Fisher Scientific
39640.18 39640.30
39640.18EA 45.6 EUR
39640.18 39640.30
Tin 99,5, mossy ACS
Tin
Purity: 99.5% CAS No.: 7440-31-5 Density: 7.310G/CC Odour: Odourless Melting Point: 231.9 deg C Boiling Point: 2270 deg C MDL No.: MFCD00133862
Formula: Sn
MW: 118.71 g/mol
Boiling Pt: 2270
Melting Pt: 232
Density: 7.31 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133862
CAS Number: 7440-31-5
EINECS: 231-141-8
Merck Index: 12,09587