Tin 99,8 Metals basis, foil, Thickness: 0.5 mm (0.02 in)
Supplier: Thermo Fisher Scientific
43234.GH 43234.HP 43234.FI
43234.GHEA 105 EUR
43234.GH 43234.HP 43234.FI
Tin 99,8 Metals basis, foil, Thickness: 0.5 mm (0.02 in)
Tin
Purity: 99.8% CAS No.: 7440-31-5 Physical Form: ≈9.1g/50x50mm Odour: Odourless MDL No.: MFCD00133862
Formula: Sn
MW: 118.71 g/mol
Boiling Pt: 2270
Melting Pt: 232
Density: 7.31 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133862
CAS Number: 7440-31-5
EINECS: 231-141-8
Merck Index: 12,09587