Tin 99,9 Metals basis, foil, Thickness: 0.18 mm (0.007 in)
Supplier: Thermo Fisher Scientific
43308.HP 43308.GH 43308.FI
43308.HPEA 632 EUR
43308.HP 43308.GH 43308.FI
Tin 99,9 Metals basis, foil, Thickness: 0.18 mm (0.007 in)
Tin
Purity: 99.9% CAS No.: 7440-31-5 Physical Form: ≈3.3g/50x50mm Odour: Odourless MDL No.: MFCD00133862
Formula: Sn
MW: 118.71 g/mol
Boiling Pt: 2270
Melting Pt: 232
Density: 7.31 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133862
CAS Number: 7440-31-5
EINECS: 231-141-8
Merck Index: 12,09587