Tin 99,995 Metals basis, slug, Puratronic®, Length: 6.35 mm (0.25 in), Ø 6.35 mm (0.25 in)
Supplier: Thermo Fisher Scientific
43416.14 43416.22
43416.14EA 102 EUR
43416.14 43416.22
Tin 99,995 Metals basis, slug, Puratronic®, Length: 6.35 mm (0.25 in), Ø 6.35 mm (0.25 in)
Tin
Purity: 99.995% CAS No.: 7440-31-5 Formula Weight: 118.710 Physical Form: ≈1.5g/slug Density: 7.310G/CC Odour: Odourless Boiling Point: 2270 deg C MDL No.: MFCD00133862
Formula: Sn
MW: 118.71 g/mol
Boiling Pt: 2270 °C
Melting Pt: 231.9 °C
Density: 7.310G/CC
Storage Temperature: Ambient
MDL Number: MFCD00133862
CAS Number: 7440-31-5
EINECS: 231-141-8
Merck Index: 12,09587