Tin ≥99% (metals basis), shot ≤ 3.175 mm (0.125 in)
Supplier: Thermo Fisher Scientific
45096.A1 45096.36 45096.22
45096.A1EA 227 EUR
45096.A1 45096.36 45096.22
Tin ≥99% (metals basis), shot ≤ 3.175 mm (0.125 in)
Tin
Purity: 99% CAS No.: 7440-31-5 Odour: Odourless MDL No.: MFCD00133862
Formula: Sn
Boiling Pt: 2270 °C (1013 hPa)
Melting Pt: 232 °C
Density: 7.31 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133862
CAS Number: 7440-31-5
EINECS: 231-141-8
Merck Index: 12,09587